Order Entry
United Kingdom
ContactUsLinkComponent
Baloxavir (Baloxavir acid), derived from the proagent Baloxavir marboxil, is a first-in-class, potent and selective cap-dependent en donuclease (CEN) inhibitor within the polymerase PA subunit of influenza A and B viruses. Baloxavir inhibits viral RN 1 * 50 mg
Catalog # MDEXHY-109025A
Supplier:  MEDCHEMEXPRESS
Baloxavir (Baloxavir acid), derived from the proagent Baloxavir marboxil, is a first-in-class, potent and selective cap-dependent en donuclease (CEN) inhibitor within the polymerase PA subunit of influenza A and B viruses. Baloxavir inhibits viral RN 1 * 50 mg
Catalog # MDEXHY-109025A
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together