Order Entry
United Kingdom
ContactUsLinkComponent
MASM7 is a mitofusin activator, and can achieve mitochondrial fusion via mitofusins. MASM7 can increase Mito AR with an EC50 value o f 75 nM in MEFs in concentration-responsively, and can promote mitochondrial fusion by directly activating MFN2 or MF 1 * 10 mg
Catalog # MDEXHY-W187305
Supplier:  MEDCHEMEXPRESS
MASM7 is a mitofusin activator, and can achieve mitochondrial fusion via mitofusins. MASM7 can increase Mito AR with an EC50 value o f 75 nM in MEFs in concentration-responsively, and can promote mitochondrial fusion by directly activating MFN2 or MF 1 * 10 mg
Catalog # MDEXHY-W187305
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together