Order Entry
United Kingdom
ContactUsLinkComponent
VTP50469 is a potent, highly selective and orally active Menin-MLL interaction inhibitor with a Ki of 104 pM. VTP50469 has potently anti-leukemia activity[1][2]. 1 * 1 mg
Catalog # MDEXHY-114162
Supplier:  MEDCHEMEXPRESS
VTP50469 is a potent, highly selective and orally active Menin-MLL interaction inhibitor with a Ki of 104 pM. VTP50469 has potently anti-leukemia activity[1][2]. 1 * 1 mg
Catalog # MDEXHY-114162
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together