Order Entry
United Kingdom
ContactUsLinkComponent
Amylin, amide, rat is a potent and high affinity ligand of Amylin receptor AMY1 and AMY3 receptors and variably of AMY2 receptors b inding studies are generally used for the latter receptor. 1 * 1 mg
Catalog # MDEXHY-P1464
Supplier:  MEDCHEMEXPRESS
Amylin, amide, rat is a potent and high affinity ligand of Amylin receptor AMY1 and AMY3 receptors and variably of AMY2 receptors b inding studies are generally used for the latter receptor. 1 * 1 mg
Catalog # MDEXHY-P1464
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together