Order Entry
United Kingdom
ContactUsLinkComponent
Relatlimab (BMS-986016) is a human monoclonal antibody anti-LAG-3 antibody generated by immunization of transgenic mice bearing huma n immunoglobulin miniloci with recombinant LAG-3 protein. Relatlimab blocks LAG-3/MHC II interaction with an IC50 val 1 * 10 mg
Catalog # MDEXHY-P99156
Supplier:  MEDCHEMEXPRESS
Relatlimab (BMS-986016) is a human monoclonal antibody anti-LAG-3 antibody generated by immunization of transgenic mice bearing huma n immunoglobulin miniloci with recombinant LAG-3 protein. Relatlimab blocks LAG-3/MHC II interaction with an IC50 val 1 * 10 mg
Catalog # MDEXHY-P99156
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together