Order Entry
United Kingdom
ContactUsLinkComponent
DS21150768 is a potent, orally active HPK1 inhibitor. DS21150768 shows potent activity to enhance T-cell function. DS21150768 has an ticancer effects[1]. 1 * 100 mg
  MDEXHY-157149
DS21150768 is a potent, orally active HPK1 inhibitor. DS21150768 shows potent activity to enhance T-cell function. DS21150768 has an ticancer effects[1]. 1 * 100 mg
  MDEXHY-157149
Frequently Bought Together