Order Entry
United Kingdom
ContactUsLinkComponent
Ruxolitinib phosphate (INCB018424 phosphate) is a potent JAK1/2 inhibitor with IC50s of 3.3 nM/2.8 nM, respectively, showing more th an 130-fold selectivity over JAK3. 1 * 100 mg
Catalog # MDEXHY-50858
Supplier:  MEDCHEMEXPRESS
Ruxolitinib phosphate (INCB018424 phosphate) is a potent JAK1/2 inhibitor with IC50s of 3.3 nM/2.8 nM, respectively, showing more th an 130-fold selectivity over JAK3. 1 * 100 mg
Catalog # MDEXHY-50858
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together