Order Entry
United Kingdom
ContactUsLinkComponent
LY379268 is a potent, selective and brain-penetrant mGlu2/3R agonist with EC50 values of 2.69 nM (mGlu2) and 4.48 nM (mGlu3). LY3792 68 has no activity on human mGlu 1a, 4a, 5a or 7a receptors. LY379268 has antioxidant and neuroprotective effects[1][ 1 * 50 mg
Catalog # MDEXHY-103558
Supplier:  MEDCHEMEXPRESS
LY379268 is a potent, selective and brain-penetrant mGlu2/3R agonist with EC50 values of 2.69 nM (mGlu2) and 4.48 nM (mGlu3). LY3792 68 has no activity on human mGlu 1a, 4a, 5a or 7a receptors. LY379268 has antioxidant and neuroprotective effects[1][ 1 * 50 mg
Catalog # MDEXHY-103558
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together