Order Entry
United Kingdom
ContactUsLinkComponent
Patritumab deruxtecan (HER3-DXd) is an antibody-drug conjugate (ADC) consisting of a fully human anti-HER3 IgG1 monoclonal antibody Patritumab (HY-P99275) attached to a topoisomerase I inhibitor payload via a tetrapeptide-based cleavable linker. Patr 1 * 5 mg
Catalog # MDEXHY-P99813
Supplier:  MEDCHEMEXPRESS
Patritumab deruxtecan (HER3-DXd) is an antibody-drug conjugate (ADC) consisting of a fully human anti-HER3 IgG1 monoclonal antibody Patritumab (HY-P99275) attached to a topoisomerase I inhibitor payload via a tetrapeptide-based cleavable linker. Patr 1 * 5 mg
Catalog # MDEXHY-P99813
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together