Order Entry
United Kingdom
ContactUsLinkComponent
Infigratinib (BGJ-398 NVP-BGJ398) is a potent inhibitor of the FGFR family with IC50s of 0.9 nM, 1.4 nM, 1 nM, and 60 nM for FGFR1, FGFR2, FGFR3, and FGFR4, respectively. 1 * 100 mg
Catalog # MDEXHY-13311
Supplier:  MEDCHEMEXPRESS
Infigratinib (BGJ-398 NVP-BGJ398) is a potent inhibitor of the FGFR family with IC50s of 0.9 nM, 1.4 nM, 1 nM, and 60 nM for FGFR1, FGFR2, FGFR3, and FGFR4, respectively. 1 * 100 mg
Catalog # MDEXHY-13311
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together