Order Entry
United Kingdom
ContactUsLinkComponent
Levofloxacin ((-)-Ofloxacin) is an orally active antibiotic and is active against both Gram-positive and Gram-negative bacteria. Lev ofloxacin inhibits the DNA gyrase and topoisomerase IV. Levofloxacin can be used for chronic periodontitis, airway i 1 * 100 mg
Catalog # MDEXHY-B0330
Supplier:  MEDCHEMEXPRESS
Levofloxacin ((-)-Ofloxacin) is an orally active antibiotic and is active against both Gram-positive and Gram-negative bacteria. Lev ofloxacin inhibits the DNA gyrase and topoisomerase IV. Levofloxacin can be used for chronic periodontitis, airway i 1 * 100 mg
Catalog # MDEXHY-B0330
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together