Order Entry
United Kingdom
ContactUsLinkComponent
TTNPB is a highly potent RAR agonist. Competitive binding assays using human RARs yield IC50s of α=5.1 nM, β= 4.5 nM, and γ=9.3 nM, respectively. 1 * 50 mg
Catalog # MDEXHY-15682
Supplier:  MEDCHEMEXPRESS
TTNPB is a highly potent RAR agonist. Competitive binding assays using human RARs yield IC50s of α=5.1 nM, β= 4.5 nM, and γ=9.3 nM, respectively. 1 * 50 mg
Catalog # MDEXHY-15682
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together