Order Entry
United Kingdom
ContactUsLinkComponent
Fasudil (HA-1077 AT877) Hydrochloride is a nonspecific RhoA/ROCK inhibitor and also has inhibitory effect on protein kinases, with an Ki of 0.33 μM for ROCK1, IC50s of 0.158 μM and 4.58 μM, 12.30 μM, 1.650 μM for ROCK2 and PKA, PKC, PKG, respectivel 1 * 200 mg
Catalog # MDEXHY-10341
Supplier:  MEDCHEMEXPRESS
Fasudil (HA-1077 AT877) Hydrochloride is a nonspecific RhoA/ROCK inhibitor and also has inhibitory effect on protein kinases, with an Ki of 0.33 μM for ROCK1, IC50s of 0.158 μM and 4.58 μM, 12.30 μM, 1.650 μM for ROCK2 and PKA, PKC, PKG, respectivel 1 * 200 mg
Catalog # MDEXHY-10341
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together