Order Entry
United Kingdom
ContactUsLinkComponent
Tucidinostat (Chidamide) is a potent and orally bioavailable HDAC enzymes class I (HDAC1/2/3) and class IIb (HDAC10) inhibitor, with IC50s of 95, 160, 67 and 78 nM, less active on HDAC8 and HDAC11 (IC50s, 733 nM, 432 nM, respectively), and shows no / 1 * 10 mg
Catalog # MDEXHY-109015
Supplier:  MEDCHEMEXPRESS
Tucidinostat (Chidamide) is a potent and orally bioavailable HDAC enzymes class I (HDAC1/2/3) and class IIb (HDAC10) inhibitor, with IC50s of 95, 160, 67 and 78 nM, less active on HDAC8 and HDAC11 (IC50s, 733 nM, 432 nM, respectively), and shows no / 1 * 10 mg
Catalog # MDEXHY-109015
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together