Order Entry
United Kingdom
ContactUsLinkComponent
CAY10566 is a potent, orally bioavailable and selective stearoyl-CoA desaturase1 (SCD1) inhibitor with IC50s of 4.5 and 26 nM in mo use and human enzymatic assays, respectively. null
Catalog # MDEXHY-15823
Supplier:  MEDCHEMEXPRESS
CAY10566 is a potent, orally bioavailable and selective stearoyl-CoA desaturase1 (SCD1) inhibitor with IC50s of 4.5 and 26 nM in mo use and human enzymatic assays, respectively. null
Catalog # MDEXHY-15823
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together