Order Entry
United Kingdom
ContactUsLinkComponent
8-Bromo-cAMP sodium (8-Br-Camp) sodium salt, a cyclic AMP analog, is an activator of cyclic AMP-dependent protein kinase (PKA). 8-Br omo-cAMP sodium salt has anti-proliferative and apoptotic effects against cancer cells[1][2]. 1 * 100 mg
Catalog # MDEXHY-12306
Supplier:  MEDCHEMEXPRESS
8-Bromo-cAMP sodium (8-Br-Camp) sodium salt, a cyclic AMP analog, is an activator of cyclic AMP-dependent protein kinase (PKA). 8-Br omo-cAMP sodium salt has anti-proliferative and apoptotic effects against cancer cells[1][2]. 1 * 100 mg
Catalog # MDEXHY-12306
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together