Order Entry
United Kingdom
ContactUsLinkComponent
Lurbinectedin (PM01183) is a DNA minor groove covalent binder with potent anti-tumour activity inhibits RMG1 and RMG2 cell growth w ith IC50 values of 1.25 and 1.16 nM, respectively. 1 * 5 mg
Catalog # MDEXHY-16293
Supplier:  MEDCHEMEXPRESS
Lurbinectedin (PM01183) is a DNA minor groove covalent binder with potent anti-tumour activity inhibits RMG1 and RMG2 cell growth w ith IC50 values of 1.25 and 1.16 nM, respectively. 1 * 5 mg
Catalog # MDEXHY-16293
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together