Order Entry
United Kingdom
ContactUsLinkComponent
2',3'-cGAMP sodium (2'-3'-cyclic GMP-AMP sodium) is a endogenous cGAMP in mammalian cells. 2',3'-cGAMP sodium binds to STING with a high affinity and is a potent inducer of interferon-β (IFNβ). 2',3'-cGAMP sodium is produced in mammalian cells in res 1 * 5 mg
Catalog # MDEXHY-100564A
Supplier:  MEDCHEMEXPRESS
2',3'-cGAMP sodium (2'-3'-cyclic GMP-AMP sodium) is a endogenous cGAMP in mammalian cells. 2',3'-cGAMP sodium binds to STING with a high affinity and is a potent inducer of interferon-β (IFNβ). 2',3'-cGAMP sodium is produced in mammalian cells in res 1 * 5 mg
Catalog # MDEXHY-100564A
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together