Order Entry
United Kingdom
ContactUsLinkComponent
Oxamic acid (oxamate) sodium salt is a lactate dehydrogenase-A (LDH-A) inhibitor. Oxamic acid sodium salt shows anti-tumor activity, and anti-proliferative activity against cancer cells, and can induce apoptosis[1][2][3]. 1 * 500 mg
Catalog # MDEXHY-W013032A
Supplier:  MEDCHEMEXPRESS
Oxamic acid (oxamate) sodium salt is a lactate dehydrogenase-A (LDH-A) inhibitor. Oxamic acid sodium salt shows anti-tumor activity, and anti-proliferative activity against cancer cells, and can induce apoptosis[1][2][3]. 1 * 500 mg
Catalog # MDEXHY-W013032A
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together