Order Entry
United Kingdom
ContactUsLinkComponent
Trolox is an analogue of vitamin E with a powerful antioxidant effect. Trolox is also a powerful inhibitor of membrane damage[1][2]. 1 * 5 g
Catalog # MDEXHY-101445
Supplier:  MEDCHEMEXPRESS
Trolox is an analogue of vitamin E with a powerful antioxidant effect. Trolox is also a powerful inhibitor of membrane damage[1][2]. 1 * 5 g
Catalog # MDEXHY-101445
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together