Order Entry
United Kingdom
ContactUsLinkComponent
Sulfasalazine (NSC 667219) is an anti-rheumatic agent for the research of rheumatoid arthritis and ulcerative colitis. Sulfasalazine can suppress NF-κB activity. Sulfasalazine is a type 1 ferroptosis inducer[1][2][3][4]. 1 * 1 g
Catalog # MDEXHY-14655
Supplier:  MEDCHEMEXPRESS
Sulfasalazine (NSC 667219) is an anti-rheumatic agent for the research of rheumatoid arthritis and ulcerative colitis. Sulfasalazine can suppress NF-κB activity. Sulfasalazine is a type 1 ferroptosis inducer[1][2][3][4]. 1 * 1 g
Catalog # MDEXHY-14655
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together