Order Entry
United Kingdom
ContactUsLinkComponent
Ganciclovir (BW 759), a nucleoside analogue, is an orally active antiviral agent with activity against CMV. Ganciclovir also has act ivity in vitro against members of the herpes group and some other DNA viruses. Ganciclovir inhibits the in vitro repl 1 * 100 mg
Catalog # MDEXHY-13637
Supplier:  MEDCHEMEXPRESS
Ganciclovir (BW 759), a nucleoside analogue, is an orally active antiviral agent with activity against CMV. Ganciclovir also has act ivity in vitro against members of the herpes group and some other DNA viruses. Ganciclovir inhibits the in vitro repl 1 * 100 mg
Catalog # MDEXHY-13637
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together