Order Entry
United Kingdom
ContactUsLinkComponent
Ciprofloxacin (Bay-09867) is a potent, orally active topoisomerase IV inhibitor. Ciprofloxacin induces mitochondrial DNA and nuclear DNA damage and lead to mitochondrial dysfunction, ROS production. Ciprofloxacin has anti-proliferative activity and i 1 * 5 g
Catalog # MDEXHY-B0356
Supplier:  MEDCHEMEXPRESS
Ciprofloxacin (Bay-09867) is a potent, orally active topoisomerase IV inhibitor. Ciprofloxacin induces mitochondrial DNA and nuclear DNA damage and lead to mitochondrial dysfunction, ROS production. Ciprofloxacin has anti-proliferative activity and i 1 * 5 g
Catalog # MDEXHY-B0356
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together