Order Entry
United Kingdom
ContactUsLinkComponent
Clavulanic acid is a naturally occurring powerful bacterial β-lactamases inhibitor for research of infections caused by bacteria, in cluding infections of the ears. Clavulanic acid is active against a wide spectrum of gram-positive and gram-negative 1 * 10 mg
Catalog # MDEXHY-A0256
Supplier:  MEDCHEMEXPRESS
Clavulanic acid is a naturally occurring powerful bacterial β-lactamases inhibitor for research of infections caused by bacteria, in cluding infections of the ears. Clavulanic acid is active against a wide spectrum of gram-positive and gram-negative 1 * 10 mg
Catalog # MDEXHY-A0256
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together