Order Entry
United Kingdom
ContactUsLinkComponent
MMAF (Monomethylauristatin F) is a potent tubulin polymerization inhibitor and is used as a antitumor agent. MMAF (Monomethylaurista tin F) is widely used as a cytotoxic component of antibody-drug conjugates (ADCs) such as vorsetuzumab mafodotin and 1 * 50 mg
Catalog # MDEXHY-15579
Supplier:  MEDCHEMEXPRESS
MMAF (Monomethylauristatin F) is a potent tubulin polymerization inhibitor and is used as a antitumor agent. MMAF (Monomethylaurista tin F) is widely used as a cytotoxic component of antibody-drug conjugates (ADCs) such as vorsetuzumab mafodotin and 1 * 50 mg
Catalog # MDEXHY-15579
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together