Order Entry
United Kingdom
ContactUsLinkComponent
Substance P (Neurokinin P) is a neuropeptide, acting as a neurotransmitter and as a neuromodulator in the CNS. The endogenous recept or for substance P is neurokinin 1 receptor (NK1-receptor, NK1R). 1 * 1 mg
Catalog # MDEXHY-P0201
Supplier:  MEDCHEMEXPRESS
Substance P (Neurokinin P) is a neuropeptide, acting as a neurotransmitter and as a neuromodulator in the CNS. The endogenous recept or for substance P is neurokinin 1 receptor (NK1-receptor, NK1R). 1 * 1 mg
Catalog # MDEXHY-P0201
Supplier:  MEDCHEMEXPRESS