Order Entry
United Kingdom
ContactUsLinkComponent
Fumarate hydratase-IN-1 (compound 2) is a cell-permeable fumarate hydratase inhibitor. Fumarate hydratase-IN-1 has antiproliferative activity against several cancer cell lines with a mean IC50 of 2.2 μM[1]. 1 * 2 mg
Catalog # MDEXHY-100004
Supplier:  MEDCHEMEXPRESS
Fumarate hydratase-IN-1 (compound 2) is a cell-permeable fumarate hydratase inhibitor. Fumarate hydratase-IN-1 has antiproliferative activity against several cancer cell lines with a mean IC50 of 2.2 μM[1]. 1 * 2 mg
Catalog # MDEXHY-100004
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together