Order Entry
United Kingdom
ContactUsLinkComponent
BAY-3827 is a potent and selective AMPK inhibitor with IC50 values of 1.4 nM at low (10 μM ATP concentration) and 15 nM at high (2 m M ATP concentration). BAY-3827 shows over 500-fold selectivity for most of the 331 kinases. BAY-3827 prevents phospho 1 * 50 mg
Catalog # MDEXHY-112083
Supplier:  MEDCHEMEXPRESS
BAY-3827 is a potent and selective AMPK inhibitor with IC50 values of 1.4 nM at low (10 μM ATP concentration) and 15 nM at high (2 m M ATP concentration). BAY-3827 shows over 500-fold selectivity for most of the 331 kinases. BAY-3827 prevents phospho 1 * 50 mg
Catalog # MDEXHY-112083
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together