Order Entry
United Kingdom
ContactUsLinkComponent
Iadademstat (ORY-1001) dihydrochloride is a selective irreversible lysine (K)-specific demethylase 1A (KDM1A/LSD1) inhibitor. 1 * 50 mg
Catalog # MDEXHY-12782T
Supplier:  MEDCHEMEXPRESS
Iadademstat (ORY-1001) dihydrochloride is a selective irreversible lysine (K)-specific demethylase 1A (KDM1A/LSD1) inhibitor. 1 * 50 mg
Catalog # MDEXHY-12782T
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together