Order Entry
United Kingdom
ContactUsLinkComponent
Caspofungin (MK-0991) diacetate is a potent antifungal agent. Caspofungin diacetate inhibits the synthesis of the fungal cell wall c omponent β-(l,3)-D-glucan[1][2]. 1 * 25 mg
Catalog # MDEXHY-17006
Supplier:  MEDCHEMEXPRESS
Caspofungin (MK-0991) diacetate is a potent antifungal agent. Caspofungin diacetate inhibits the synthesis of the fungal cell wall c omponent β-(l,3)-D-glucan[1][2]. 1 * 25 mg
Catalog # MDEXHY-17006
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together