Order Entry
United Kingdom
ContactUsLinkComponent
CGP37157 is a potent, selective inhibitor of Na+/Ca2+ exchanger, inhibiting the Na+-induced Ca2+-release from guinea-pig heart mitoc hondria, with an IC50 of 0.8 μM. 1 * 5 mg
Catalog # MDEXHY-15754
Supplier:  MEDCHEMEXPRESS
CGP37157 is a potent, selective inhibitor of Na+/Ca2+ exchanger, inhibiting the Na+-induced Ca2+-release from guinea-pig heart mitoc hondria, with an IC50 of 0.8 μM. 1 * 5 mg
Catalog # MDEXHY-15754
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together