Order Entry
United Kingdom
ContactUsLinkComponent
Mavacamten (MYK461) is an orally active modulator of cardiac myosin, with IC50s of 490, 711 nM for bovine cardiac and human cardiac, respectively. 1 * 10 mg
Catalog # MDEXHY-109037
Supplier:  MEDCHEMEXPRESS
Mavacamten (MYK461) is an orally active modulator of cardiac myosin, with IC50s of 490, 711 nM for bovine cardiac and human cardiac, respectively. 1 * 10 mg
Catalog # MDEXHY-109037
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together