Order Entry
United Kingdom
ContactUsLinkComponent
AZD-5991 is a potent and selective Mcl-1 inhibitor with an IC50 of 0.7 nM in FRET assay and a Kd of 0.17 nM in surface plasmon reson ance (SPR) assay[1]. 1 * 50 mg
Catalog # MDEXHY-101533
Supplier:  MEDCHEMEXPRESS
AZD-5991 is a potent and selective Mcl-1 inhibitor with an IC50 of 0.7 nM in FRET assay and a Kd of 0.17 nM in surface plasmon reson ance (SPR) assay[1]. 1 * 50 mg
Catalog # MDEXHY-101533
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together