Order Entry
United Kingdom
ContactUsLinkComponent
Busulfan is a potent alkylating antineoplastic agent. Busulfan causes DNA damage by cross-linking DNAs and DNA and proteins. Busulfa n inhibits thioredoxin reductase. Busulfan induces apoptosis. Busulfan is an immunosuppressive and myeloablative chem 1 * 5 g
Catalog # MDEXHY-B0245
Supplier:  MEDCHEMEXPRESS
Busulfan is a potent alkylating antineoplastic agent. Busulfan causes DNA damage by cross-linking DNAs and DNA and proteins. Busulfa n inhibits thioredoxin reductase. Busulfan induces apoptosis. Busulfan is an immunosuppressive and myeloablative chem 1 * 5 g
Catalog # MDEXHY-B0245
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together