Order Entry
United Kingdom
ContactUsLinkComponent
Proteinase K (Protease K) is a nonspecific serine protease that is useful for general digestion of proteins. Proteinase K is active in the presence of SDS or urea and over a wide range of pH (4-12), salt concentrations, and temperatures. Proteinase K 1 * 25 mg
Catalog # MDEXHY-108717
Supplier:  MEDCHEMEXPRESS
Proteinase K (Protease K) is a nonspecific serine protease that is useful for general digestion of proteins. Proteinase K is active in the presence of SDS or urea and over a wide range of pH (4-12), salt concentrations, and temperatures. Proteinase K 1 * 25 mg
Catalog # MDEXHY-108717
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together