Order Entry
United Kingdom
ContactUsLinkComponent
Asunaprevir (BMS-650032) is a potent and orally bioavailable hepatitis C virus (HCV) NS3 protease inhibitor, with IC50 of 0.2 nM-3.5 nM[1]. Asunaprevir inhibits SARS-CoV-2 3CLpro activity[5]. 1 * 5 mg
Catalog # MDEXHY-14434
Supplier:  MEDCHEMEXPRESS
Asunaprevir (BMS-650032) is a potent and orally bioavailable hepatitis C virus (HCV) NS3 protease inhibitor, with IC50 of 0.2 nM-3.5 nM[1]. Asunaprevir inhibits SARS-CoV-2 3CLpro activity[5]. 1 * 5 mg
Catalog # MDEXHY-14434
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together