Order Entry
United Kingdom
ContactUsLinkComponent
4-Hydroperoxy cyclophosphamide is the active metabolite form of the precursor Cyclophosphamide. 4-Hydroperoxy cyclophosphamide cross -links DNA to induce T cell apoptosis independent of caspase receptor activation, and can activate the mitochondrial 1 * 10 mg
  MDEXHY-117433
4-Hydroperoxy cyclophosphamide is the active metabolite form of the precursor Cyclophosphamide. 4-Hydroperoxy cyclophosphamide cross -links DNA to induce T cell apoptosis independent of caspase receptor activation, and can activate the mitochondrial 1 * 10 mg
  MDEXHY-117433
Frequently Bought Together