Order Entry
United Kingdom
ContactUsLinkComponent
Osunprotafib (ABBV-CLS-484) is a potent PTPN1 and PTPN2 inhibitor with subnanomolar activity. Osunprotafib has antitumor activity, e nhances the immune response and increases the sensitivity of tumors to immune-mediated killing[1]. 1 * 1 mg
Catalog # MDEXHY-145923
Supplier:  MEDCHEMEXPRESS
Osunprotafib (ABBV-CLS-484) is a potent PTPN1 and PTPN2 inhibitor with subnanomolar activity. Osunprotafib has antitumor activity, e nhances the immune response and increases the sensitivity of tumors to immune-mediated killing[1]. 1 * 1 mg
Catalog # MDEXHY-145923
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together