Order Entry
United Kingdom
ContactUsLinkComponent
U0126 is a potent, non-ATP competitive and selective MEK1 and MEK2 inhibitor, with IC50s of 72 nM and 58 nM, respectively. U0126 is an autophagy and mitophagy inhibitor[1][2][3][4]. 1 * 25 mg
Catalog # MDEXHY-12031A
Supplier:  MEDCHEMEXPRESS
U0126 is a potent, non-ATP competitive and selective MEK1 and MEK2 inhibitor, with IC50s of 72 nM and 58 nM, respectively. U0126 is an autophagy and mitophagy inhibitor[1][2][3][4]. 1 * 25 mg
Catalog # MDEXHY-12031A
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together