Order Entry
United Kingdom
ContactUsLinkComponent
Palbociclib (PD 0332991) isethionate is an orally active selective CDK4 and CDK6 inhibitor with IC50 values of 11 and 16 nM, respect ively. Palbociclib isethionate has potent anti-proliferative activity and induces cell cycle arrest in cancer cells, 1 * 100 mg
Catalog # MDEXHY-A0065
Supplier:  MEDCHEMEXPRESS
Palbociclib (PD 0332991) isethionate is an orally active selective CDK4 and CDK6 inhibitor with IC50 values of 11 and 16 nM, respect ively. Palbociclib isethionate has potent anti-proliferative activity and induces cell cycle arrest in cancer cells, 1 * 100 mg
Catalog # MDEXHY-A0065
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together