Order Entry
United Kingdom
ContactUsLinkComponent
ISRIB (trans-isomer) is a potent inhibitor of PERK with an IC50 of 5 nM. ISRIB potently reverses the effects of eIF2α phosphorylatio n (IC50=5 nM). 1 * 10 mg
Catalog # MDEXHY-12495
Supplier:  MEDCHEMEXPRESS
ISRIB (trans-isomer) is a potent inhibitor of PERK with an IC50 of 5 nM. ISRIB potently reverses the effects of eIF2α phosphorylatio n (IC50=5 nM). 1 * 10 mg
Catalog # MDEXHY-12495
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together