Order Entry
United Kingdom
ContactUsLinkComponent
diABZI STING agonist-1 (trihydrochloride) is a selective stimulator of interferon genes (STING) receptor agonist, with EC50s of 130 , 186 nM for human and mouse, respectively. 1 * 1 mg
Catalog # MDEXHY-112921B
Supplier:  MEDCHEMEXPRESS
diABZI STING agonist-1 (trihydrochloride) is a selective stimulator of interferon genes (STING) receptor agonist, with EC50s of 130 , 186 nM for human and mouse, respectively. 1 * 1 mg
Catalog # MDEXHY-112921B
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together