Order Entry
United Kingdom
ContactUsLinkComponent
DT2216 is a potent and selective BCL-XL (Bcl-2 family member) degrader based on PROTAC technology. DT2216 causes effective degradati on of BCL-XL protein by recruiting Von Hippel-Lindau (VHL) E3 ubiquitin ligase. DT2216 inhibits various BCL-XL-depend 1 * 25 mg
Catalog # MDEXHY-130604
Supplier:  MEDCHEMEXPRESS
DT2216 is a potent and selective BCL-XL (Bcl-2 family member) degrader based on PROTAC technology. DT2216 causes effective degradati on of BCL-XL protein by recruiting Von Hippel-Lindau (VHL) E3 ubiquitin ligase. DT2216 inhibits various BCL-XL-depend 1 * 25 mg
Catalog # MDEXHY-130604
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together