Order Entry
United Kingdom
ContactUsLinkComponent
H-89 is a potent and selective inhibitor of cyclic AMP-dependent protein kinase (protein kinase A) with IC50 of 48 nM and has weak i nhibition on PKG, PKC, Casein Kinase, and others kinases. 1 * 100 mg
Catalog # MDEXHY-15979
Supplier:  MEDCHEMEXPRESS
H-89 is a potent and selective inhibitor of cyclic AMP-dependent protein kinase (protein kinase A) with IC50 of 48 nM and has weak i nhibition on PKG, PKC, Casein Kinase, and others kinases. 1 * 100 mg
Catalog # MDEXHY-15979
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together