Order Entry
United Kingdom
ContactUsLinkComponent
Linsitinib (OSI-906) is a potent, selective and orally bioavailable dual inhibitor of the IGF-1 receptor and insulin receptor (IR) w ith IC50s of 35 and 75 nM, respectively[1]. 1 * 5 mg
Catalog # MDEXHY-10191
Supplier:  MEDCHEMEXPRESS
Linsitinib (OSI-906) is a potent, selective and orally bioavailable dual inhibitor of the IGF-1 receptor and insulin receptor (IR) w ith IC50s of 35 and 75 nM, respectively[1]. 1 * 5 mg
Catalog # MDEXHY-10191
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together