Order Entry
United Kingdom
ContactUsLinkComponent
Favipiravir (T-705) is a potent viral RNA polymerase inhibitor, it is phosphoribosylated by cellular enzymes to its active form, Fav ipiravir-ribofuranosyl-5′-triphosphate (RTP). Favipiravir-RTP inhibits the influenza viral RNA-dependent RNA polymera 1 * 5 mg
Catalog # MDEXHY-14768
Supplier:  MEDCHEMEXPRESS
Favipiravir (T-705) is a potent viral RNA polymerase inhibitor, it is phosphoribosylated by cellular enzymes to its active form, Fav ipiravir-ribofuranosyl-5′-triphosphate (RTP). Favipiravir-RTP inhibits the influenza viral RNA-dependent RNA polymera 1 * 5 mg
Catalog # MDEXHY-14768
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together