Order Entry
United Kingdom
ContactUsLinkComponent
Indisulam (E 7070) is a carbonic anhydrase inhibitor with anticancer activity. Indisulam (E 7070) is a sulfonamide agent that target s the G1 phase of the cell cycle. Indisulam (E 7070) causes a blockade in the G1/S transition through inhibition of t 1 * 1 mg
Catalog # MDEXHY-13650
Supplier:  MEDCHEMEXPRESS
Indisulam (E 7070) is a carbonic anhydrase inhibitor with anticancer activity. Indisulam (E 7070) is a sulfonamide agent that target s the G1 phase of the cell cycle. Indisulam (E 7070) causes a blockade in the G1/S transition through inhibition of t 1 * 1 mg
Catalog # MDEXHY-13650
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together