Order Entry
United Kingdom
ContactUsLinkComponent
Imiquimod (R 837), an immune response modifier, is a selective toll like receptor 7 (TLR7) agonist. Imiquimod exhibits antiviral and antitumor effects in vivo. Imiquimod can be used for the research of external genital, perianal warts, cancer and CO 1 * 200 mg
Catalog # MDEXHY-B0180
Supplier:  MEDCHEMEXPRESS
Imiquimod (R 837), an immune response modifier, is a selective toll like receptor 7 (TLR7) agonist. Imiquimod exhibits antiviral and antitumor effects in vivo. Imiquimod can be used for the research of external genital, perianal warts, cancer and CO 1 * 200 mg
Catalog # MDEXHY-B0180
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together