Order Entry
United Kingdom
ContactUsLinkComponent
Deferoxamine (Deferoxamine B) is an iron chelator (binds to Fe(III) and many other metal cations), is widely used to reduce iron acc umulation and deposition in tissues. Deferoxamine upregulates HIF-1α levels with good antioxidant activity. Deferoxam 1 * 5 mg
Catalog # MDEXHY-B1625
Supplier:  MEDCHEMEXPRESS
Deferoxamine (Deferoxamine B) is an iron chelator (binds to Fe(III) and many other metal cations), is widely used to reduce iron acc umulation and deposition in tissues. Deferoxamine upregulates HIF-1α levels with good antioxidant activity. Deferoxam 1 * 5 mg
Catalog # MDEXHY-B1625
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together