Order Entry
United Kingdom
ContactUsLinkComponent
G6PDi-1 is a reversible and non-competitive glucose-6-phosphate dehydrogenase (G6PD) inhibitor with an IC50 of 0.07 μM for human G6P D. G6PDi-1 depletes NADPH most strongly in lymphocytes. G6PDi-1 markedly decreases inflammatory cytokine production i 1 * 5 mg
Catalog # MDEXHY-W107464
Supplier:  MEDCHEMEXPRESS
G6PDi-1 is a reversible and non-competitive glucose-6-phosphate dehydrogenase (G6PD) inhibitor with an IC50 of 0.07 μM for human G6P D. G6PDi-1 depletes NADPH most strongly in lymphocytes. G6PDi-1 markedly decreases inflammatory cytokine production i 1 * 5 mg
Catalog # MDEXHY-W107464
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together